Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-Tris Hydrochloride, Biological Buffer, 99%, Spectrum™ Chemical
SDP

Catalog No. 18609132 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
10 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-132 25 g Poly Bottle
18-609-134 500 g Poly Bottle
18-609-135 10 kg Poly Bottle
18-609-136 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-132 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B311225GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 124763-51-5
Molecular Formula C8H20ClNO5
Molecular Weight (g/mol) 245.70
InChI Key VEYRVLHAMHQVTC-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol hydrochloride
SMILES Cl.OCCN(CCO)C(CO)(CO)CO

Specifications

CAS Max % 1
Physical Form Crystalline Powder
Quantity 25 g
Packaging Poly Bottle
Percent Purity 99%
Grade Reagent
Chemical Name or Material Bis-Tris Hydrochloride, Biological Buffer

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.