Learn More
Abcam Bis-Tris Propane Buffer, >99% Purity, 50 g
Description
- Molecular Weight: 282.34 g/mol
- Purity: >99%
- Buffering Range: pH 6.3 to 9.5
- pKa Values: 6.8 (pKa¹) and 9.0 (pKa²)
- Composition: 100% Bis-Tris propane
- Applications: Used to enhance stability or activity of restriction enzymes
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Powder |
| Buffer | 100% Bis-Tris propane |
| Content And Storage | Store at ambient temperature. |
| Shipping Condition | Ambient |
| Formulation | 100% Bis-Tris propane |
| Quantity | 50 g |
| For Use With (Application) | Restriction enzyme stability, restriction enzyme activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.