Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisacodyl 98.0+%, TCI America™
SDP

Catalog No. B506625G Shop All Tokyo Chemical Industry (TCI) Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B506625G 25 g
B50665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B506625G Supplier TCI America Supplier No. B506625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 603-50-9
Molecular Formula C22H19NO4
Molecular Weight (g/mol) 361.40
MDL Number MFCD00038039
InChI Key KHOITXIGCFIULA-UHFFFAOYSA-N
Synonym bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
PubChem CID 2391
IUPAC Name 4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 361.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bisacodyl
EINECSNumber (9)-2202
RTECSNumber SM8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.