Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bisbenzimide H 33342, DNA Fluorescent Stain, Solid, 25 mg

Catalog No. 502583250
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3250 Supplier Abcam Supplier No. AB14559725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable, adenine-thymine selective DNA fluorescent stain that labels DNA, chromosomes, and nuclei.

  • Purity: >98%
  • Molecular Weight: 561.9 Da
  • Form: Solid
  • Applications: Cell-permeable, adenine-thymine selective DNA fluorescent stain; labels DNA, chromosomes, and nuclei
  • Mechanism: DNA minor groove-directed agent; stimulates topoisomerase I-mediated DNA cleavage
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule DNA
Substrate Properties Chemical Substrate
Substrate DNA
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Detection Method Fluorescence
Formulation 100% 2'-(4-ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzimidazole
Quantity 25 mg
For Use With (Application) Labeling DNA, chromosomes and nuclei
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.