Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Bisbenzimide H 33342 Fluorochrome, Trihydrochloride Calbiochem™,

Catalog No. 382065100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-206-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-206-5100MG Supplier MilliporeSigma™ Supplier No. 382065100MG
Add to Cart
Add to Cart

Cell-permeable, adenine-thymine-specific fluorescent stain

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23491-52-3
Molecular Formula C27H28N6O
Molecular Weight (g/mol) 452.562
InChI Key PRDFBSVERLRRMY-UHFFFAOYSA-N
Synonym hoechst 33342, bisbenzimide, hoechst33342, unii-99kzs6cnzx, 99kzs6cnzx, 2'-4-ethoxyphenyl-5-4-methylpiperazin-1-yl-1h,1'h-2,5'-bibenzo d imidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-benzimidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-1h-benzimidazole, 2'-4'-ethoxyphenyl-5-4-methylpiperazin-1-yl-2,5'-bis-1h-benzimidazole trihydrochloride trihydrate, 2'-p-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bibenzimidazole
PubChem CID 1464
ChEBI CHEBI:51232
IUPAC Name 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazole
SMILES CCOC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N6CCN(CC6)C

Specifications

Color Yellow
Quantity 100 mg
UN Number UN 2811
Solubility Information Easily soluble in cold water
Formula Weight 561.9
Percent Purity ≥95%
Physical Form Solid
Chemical Name or Material Bisbenzimide H 33342 Fluorochrome
DOTInformation DOT Class: 6.1, Packaging Group: III
RTECSNumber DT4200000
Recommended Storage 2° to 8°C (35.6° to 46.4°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.