Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bisindolylmaleimide XI Hydrochloride, PKC Inhibitor, Solid, 1 mg

Catalog No. 502706086
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6086 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6086 Supplier Abcam Supplier No. AB1437831MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Selective, cell-permeable protein kinase C inhibitor with high potency for PKCα and PKCβI.

  • Molecular Weight: 489 Da
  • Purity: >98%
  • Form: Solid
  • Inhibitor Activity: Selective, cell-permeable PKC inhibitor; displays 10-fold greater selectivity for PKCα (IC50 = 9 nM) and 4-fold greater selectivity for PKCβI (IC50 = 28 nM) over Ca²+-independent PKCε (IC50 = 108 nM).
  • Additional Targets: Inhibits GRK2, GRK5, and GRK6 (IC50 values are 29, 3.6, and 16 μM, respectively).
  • Biological Effects: Enhances cardiac contractility and protection from heart failure; prevents T-cell activation and proliferation (IC50s ranging from 30-150 nM).
  • CAS Number: 151342-35-7

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Protein Kinase C (PKC), GRK2, GRK5, GRK6
Shipping Condition Ambient
Alternate Part Number Ro 32-0432 hydrochloride
Buffer 100% Ro 32-0432 hydrochloride
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Ro 32-0432 hydrochloride
Quantity 1 mg
For Use With (Application) Protein kinase C inhibition, T-cell activation inhibition, Cardiac contractility research, Chronic inflammatory response research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.