Learn More
SPEX CERTIPREP LLC 3P BISMUTH 100 PPM. 2% NTRC 250ML
Supplier: SPEX CERTIPREP LLC 3P 10061250
(HPS by Zeptometrix) 100 ug/mL Bismuth in 2% HNO3 Exp Date: 12 months This standard is traceable to NIST and certified Guide 34 and 17025
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.