Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Carbonate, MP Biomedicals

Catalog No. ICN213703 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213703 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213703 Supplier MP Biomedicals, Inc Supplier No. 0521370380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O

Specifications

Quantity 100 g
Formula Weight 510.01
Chemical Name or Material Bismuth Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.