accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBiOOOBiO

CAS RN 5892-10-4

IUPAC Name: ({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
Synonyms: bismuth subcarbonate dibismuth carbonate dioxide
Molecular Weight (g/mol): 509.97
Molecular Formula: CBi2O5
InChi Key: MGLUJXPJRXTKJM-UHFFFAOYSA-L
SMILES: O=[Bi]OC(=O)O[Bi]=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)