Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Citrate, Powder, 50.0-52.5%, Spectrum™ Chemical
SDP

Catalog No. 18609094 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-094 25 g Poly Pail
18-609-093 125 g Poly Pail
18-609-096 500 g Poly Pail
18-609-092 12 kg Poly Pail
18-609-095 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-094 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B110125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 813-93-4

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 25 g
Percent Purity 50.0 to 52.5%
Grade Ungraded
Packaging Poly Pail
Chemical Name or Material Bismuth Citrate, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.