Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) fluoride, anhydrous, 99%

Catalog No. AA1307522
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1307522 100 g
AA1307514 25 g
AA1307536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1307522 Supplier Thermo Scientific Chemicals Supplier No. 01307522
Only null left
Add to Cart

Manufacturing of bismuth pentafluoride | CAS: 7787-61-3 | BiF3 | 265.976 g/mol

It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-61-3
Molecular Formula BiF3
Molecular Weight (g/mol) 265.976
MDL Number MFCD00010893
InChI Key BRCWHGIUHLWZBK-UHFFFAOYSA-K
Synonym bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
PubChem CID 82233
IUPAC Name trifluorobismuthane
SMILES F[Bi](F)F

Specifications

Melting Point 727°C
Physical Form Powder
Quantity 100 g
UN Number UN3260
Merck Index 14,1265
Solubility Information Insoluble in water; Soluble in HF.
Formula Weight 265.98
Percent Purity 99%
Odor Odorless
Refractive Index 1.74
Sensitivity Hygroscopic
Chemical Name or Material Bismuth(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-124-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.