Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) trifluoromethanesulfonate, 99%

Catalog No. AAL1968706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1968706 5 g
AAL1968714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1968706 Supplier Thermo Scientific Chemicals Supplier No. L1968706
  • $97.80
    $83.65 / Each of 1

    Save $14.15

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 88189-03-1 | C3BiF9O9S3 | 656.17 g/mol

Bismuth(III) trifluoromethanesulfonate acts as a catalyst in Friedel-Crafts acylation and cycloisomerization of allene-enol ethers. It behaves as a direct substitution catalyst and involved in the substitution of allylic, propargylic, and benzylic alcohols with sulfonamides, carboxamides and carbamates. Further, it is also used in Mukaiyama aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) trifluoromethanesulfonate acts as a catalyst in Friedel-Crafts acylation and cycloisomerization of allene-enol ethers. It behaves as a direct substitution catalyst and involved in the substitution of allylic, propargylic, and benzylic alcohols with sulfonamides, carboxamides and carbamates. Further, it is also used in Mukaiyama aldol reactions.

Solubility
Soluble in organics acetonitrile, dioxane, dimethyl formamide and dimethyl sulfoxide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88189-03-1
Molecular Formula C3BiF9O9S3
Molecular Weight (g/mol) 656.17
MDL Number MFCD02093669
InChI Key NYENCOMLZDQKNH-UHFFFAOYSA-K
Synonym bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
PubChem CID 9917655
IUPAC Name bismuth;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]

Specifications

Melting Point >300°C
Linear Formula (CF3SO3)3Bi
Quantity 5 g
UN Number UN3261
Sensitivity Hygroscopic
Solubility Information Soluble in organics acetonitrile,dioxane,dimethyl formamide and dimethyl sulfoxide.
Formula Weight 656.19
Percent Purity 99%
Chemical Name or Material Bismuth(III) trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.