Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Nitrate, 0.05 M, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88505 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88505 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88505 Supplier Aqua Phoenix Scientific Supplier No. BN6905P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12027-67-7 , 7732-18-5 , 7697-37-2
Molecular Formula H24Mo7N6O24
Molecular Weight (g/mol) 1163.85
MDL Number MFCD00064633
InChI Key MISTUIVWPMGXFX-UHFFFAOYSA-T
IUPAC Name hexaammonium dodecaoxododecacyclo[7.5.1.1³,⁵.1³,¹³.1⁵,⁷.1⁷,⁹.0¹,⁴.0¹,⁶.0¹,⁸.0²,¹³.0¹¹,¹⁴.0¹¹,¹⁵]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium)
SMILES [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11

Specifications

CAS Max % <100%
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.05 M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.