Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subcarbonate, USP, 97.6-100.7%, Spectrum™ Chemical
SDP

Catalog No. 18601005 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
12 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-005 12 kg Amber Glass Bottle
18-601-002 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-005 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B196912KGBL

missing translation for 'validMainframeMsg'

B1969, 5892-10-4, Approx. (BiO)2CO3

Spectrum™ Chemical Bismuth Subcarbonate, USP is used as a filler in radiopaque catheters which can be seen by x-ray. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
IUPAC Name ({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
SMILES O=[Bi]OC(=O)O[Bi]=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Percent Purity 97.6 to 100.7%
Grade USP
Loss on Drying 1%
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Subsalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.