Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subgallate, USP, 52-57%, Spectrum™ Chemical
SDP

Catalog No. 18601009 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-009 125 g Amber Glass Bottle
18-601-010 500 g Amber Glass Bottle
18-601-011 2.5 kg Amber Glass Bottle
18-601-012 12 kg Amber Glass Bottle
18-601-013 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-009 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1116125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

B1116, 99-26-3, C7H5BiO6

Spectrum™ Chemical Bismuth Subgallate, USP is used to treat malodor by deodorizing flatulence and stool odors. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 99-26-3
Molecular Formula C7H5BiO6
Molecular Weight (g/mol) 394.09
MDL Number MFCD00044980
InChI Key JAONZGLTYYUPCT-UHFFFAOYSA-K
IUPAC Name 2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
SMILES O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O

Specifications

Packaging Amber Glass Bottle
CAS Max % 100%
Quantity 125 g
Percent Purity 52 to 57%
Grade USP
Chemical Name or Material Bismuth Subgallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.