Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Bismuth Subgallate USP
SDP

Catalog No. 14186237 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-237 100 g
14-186-158 25 g
14-185-494 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-186-237 Supplier Medisca Supplier No. 104605
Add to Cart
Add to Cart

Bismuth Subgallate USP is an amorphous, bright yellow powder that is odorless. It is stable in air, but affected by light.

Bismuth Subgallate USP is an amorphous, bright yellow powder that is odorless. It is stable in air, but affected by light.

Alternative name(s): Gallic acid bismuth basic salt

Specifications

Chemical Name or Material Bismuth Subgallate USP
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 99-26-3
Type Drug Standard
Molecular Weight (g/mol) 394.091
Quantity 100 g
Percent Purity 52 to 57 % as Bi2O3
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.