Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subnitrate, Powder, USP, 79%, Spectrum™ Chemical
SDP

Catalog No. 18601016 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-016 2.5 kg Amber Glass Bottle
18-601-017 12 kg Poly Pail
18-601-018 50 kg Fiber Drum
18-601-014 125 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-016 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BI1502.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

BI150, 1304-85-4, Bi5O(OH)9(NO3)4

Spectrum™ Chemical Bismuth Subnitrate, Powder, USP is used as a dermatological agent and antidiarrheal. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1304-85-4
Molecular Formula BiN3O9
Molecular Weight (g/mol) 394.99
InChI Key PPNKDDZCLDMRHS-UHFFFAOYSA-N
IUPAC Name bis(nitrooxy)bismuthanyl nitrate
SMILES [O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
UN Number UN1477
Percent Purity 79%
Grade USP
Loss on Drying 3%
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.