Learn More
Description
Bismuth Subnitrate USP is a white, slightly hygroscopic powder.
Alternative name(s): Bismuth hydroxide nitrate oxide
Specifications
Specifications
| Chemical Name or Material | Bismuth Subnitrate USP |
| Packaging | Bottle, 30mL, Amber Glass, Wide Mouth Packer, 28-400 |
| CAS | 1304-85-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 1461.99 |
| Quantity | 25 g |
| Percent Purity | ≥79.0 % of Bi2O3 |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
