Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Bismuth Subnitrate USP
SDP

Catalog No. 14185590 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-590 100 g
14-185-474 25 g
14-185-585 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-185-590 Supplier Medisca Supplier No. 104705
Add to Cart
Add to Cart

Bismuth Subnitrate USP is a white, slightly hygroscopic powder. Alternative name(s): Bismuth hydroxide nitrate oxide

Bismuth Subnitrate USP is a white, slightly hygroscopic powder.

Alternative name(s): Bismuth hydroxide nitrate oxide

Specifications

Chemical Name or Material Bismuth Subnitrate USP
Packaging Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 1304-85-4
Type Drug Standard
Molecular Weight (g/mol) 1461.99
Quantity 100 g
Percent Purity ≥79.0 % of Bi2O3
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.