Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bisphenol A Recombinant Mouse Monoclonal Antibody (2H3)

Catalog No. MA566611
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566611 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566611 Supplier Invitrogen™ Supplier No. MA566611
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Bisphenol A Recombinant Monoclonal Antibody for ELISA

Bisphenol A (BPA) is predominantly used in the production of polycarbonate plastics and epoxy resins that are used in many products. It is released into the environment and food. BPA is an endocrine disruptor with estrogenic and obesogenic properties. It influences reproduction and has an epigenetic effect already in the foetus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bisphenol A
Applications ELISA
Classification Recombinant Monoclonal
Clone 2H3
Concentration 10.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Conjugate of BPA hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.