Learn More
Medchemexpress LLC Bisphenol B-d8 | 2469735-90-6 | 98.9% | 250.36 | C16H10D8O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW6539755MG
Encompass_Preferred
Bisphenol B-d8 is a deuterium-labeled isotopologue of bisphenol B provided in milligram quantities for analytical and research use. It is intended as a stable isotope internal standard for mass spectrometry and quantitative analysis of bisphenol compounds.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • Available in small milligram packs suitable for analytical workflows.
  • High purity, 98.9%, suitable for analytical applications.
  • Chemical formula C16H10D8O2 and molecular weight 250.36.
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-83936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.