Learn More
Abcam Bisulfite Conversion Kit, Whole Cell, 80 Test
Description
- Assay time: 3 hr
- Sample types: Saliva, Urine, Tissue, Suspension cells, Adherent cells
- Citation count: 1
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | DNA |
| Format | Kit |
| No. of Reactions | 80 Tests |
| Detection Location | In-Solution Detection |
| Sample Type | Saliva, Urine, Tissue, Suspension Cells, Adherent Cells |
| Quantity | 80 Tests |
| For Use With (Application) | Bisulfite conversion |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.