Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ BiTek™ Proteose Peptone
GREENER_CHOICE

Catalog No. 253310TF Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-331-0TF 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-331-0TF Supplier Gibco™ Supplier No. 253310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Developed as an alternative to Gibco Bacto Proteose Peptones for scale-up to production applications.

Enzymatic digests of protein. Gibco BiTek Proteose Peptone was developed to provide an alternative product to Bacto™ Proteose Peptone, with similar growth characteristics.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Peptone
For Use With (Application) Scale-up to production applications
Product Line BiTek
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.