Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biuret Reagent TS, Ricca Chemical
SDP

Catalog No. 102525 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
102525 10 L Cubitainer
R1025000100 100 mL Natural Poly Bottle
1025-16 500 mL Natural Poly Bottle
1025-32 1 L Natural Poly Bottle
10251 4 L Natural Poly Bottle
10255 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 102525 Supplier Ricca Chemical Company Supplier No. 10252.5
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Biuret Reagent
CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
Grade USP
Packaging Cubitainer
pH >12
Quantity 10 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.