Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIX NHE1 inhibitor
GREENER_CHOICE

Catalog No. p-7110838
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-121-0 10 mg
55-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 55-121-0 Supplier Tocris Bioscience™ Supplier No. 5512/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NHE1 inhibitor

BIX NHE1 inhibitor is a potent and selective NHE1 inhibitor (IC50 = 31 nM). Selective for NHE1 over NHE2 and NHE3. Prevents ischemic damage in an ischemia reperfusion injury isolated rat heart model ex vivo. Prevents phenylephrine -induced cardiomyocyte hypertrophy in vitro, and attenuates cardiac hypertrophy and left ventricular dysfunction postinfarction in rats. Orally bioavailable.

Specifications

Chemical Name or Material 4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide hydrochloride
CAS 1422252-46-7
Quantity 10 mg
Target Na+/Ca2+ Exchanger Inhibitors
Molecular Formula C16H19F3N4O2.HCl
Formula Weight 392.8
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.