Learn More
Cygnus Technologies Cygnus Technologies BL21(DE3) HCP ELISA Kit, Detects BL21(DE3) HCP Impurities, High Specificity and Sensitivity, 96 Well Plate
Supplier:  Cygnus Technologies F1060
Encompass
The Cygnus Technologies BL21(DE3) Host Cell Protein ELISA Kit is designed for precise detection of *E. coli* host cell protein impurities in biopharmaceuticals. Its high specificity and sensitivity aid in regulatory compliance and optimization of purification processes. With a broad antibody reactivity, it detects a wide range of HCPs, setting it apart from other assays. This kit ensures reliable quality control and product release testing, making it essential for researchers and manufacturers alike.
Catalog No. NC2881653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.