Learn More
McMaster-Carr BLACK DELRIN ACETAL RESIN SH
Supplier:  McMaster-Carr 8575K216
Encompass
Delrin acetal resin tubes from McMaster-Carr are made from wear-resistant, easy-to-machine Delrin plastic. They have excellent sliding properties and low friction, making them suitable for use in bearings, rollers, and bushings. The tubes have precise dimensions and tight tolerances. They are available in standard sizes with outside diameters from 1/4" to 2" and wall thicknesses from 1/32" to 1/4". The tubes can be cut to length and machined. Delrin offers good strength and stiffness. It resists fuels, solvents, and acids. The maximum service temperature is 180°F.
Catalog No. NC2603959
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.