Learn More
Supertek™ Black Enamel Hooked Weights
GREENER_CHOICESCI_ED
Catalog No. S160126 Shop All Supertek Scientific Products
Change view
Click to view available options
Weight (Metric):
10 g
10, 50, 100, 500, 1000 g, 20 g and 200 g
100 g
1000 g
20 g
200 g
50 g
500 g
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
S160126 200 g
S160121 10, 50, 100, 500, 1000 g, 20 g and 200 g
S160122 10 g
S160123 100 g
S160124 1000 g
S160125 20 g
S160127 50 g
S160128 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S160126 Supplier Supertek™ Supplier No. PH10185/200
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The steel black enamel coated hooked weights are designed to link together via their hooks. The bottoms of the hooks are recessed and contain a bar, enabling weights to be hung from each other.
  • Non-movable hook on the top
  • Available in a set of 9 in a molded container, which includes 1 each of 10 g, 50 g, 100 g, 500 g, and 1000 g, and 2 each of 20 g and 200 g
  • All weights can also be sold individually
Specifications
Product Type Hook Weight
Weight (Metric) 200 g