Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps

Catalog No. 02993291
Change view
Click to view available options
Neck Finish:
33-400
38-400
43-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Packaging:
Bagged
Bulk
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Packaging
02-993-291 83-400 Bagged
02-993-332 33-400 Bagged
02-991-187 38-400 Bulk
02-993-344 48-400 Bagged
02-991-158 48-400 Bulk
02-993-313 38-400 Bagged
02-991-016 89-400 Bulk
02-993-363 70-400 Bagged
02-993-364 58-400 Bagged
02-991-025 83-400 Bulk
02-993-362 63-400 Bagged
02-993-350 53-400 Bagged
02-993-339 43-400 Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 02-993-291 Supplier Qorpak™ Supplier No. CAP00084
Add to Cart
Add to Cart

Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.

  • Paperless composition avoids contamination from fiber or paper particles
  • For non-oil products filled at room temperature

Specifications

Color Black
Material Phenolic Resin
No. per Case 144
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material Polyethylene
Neck Finish 83-400
Disposable Reusable
Product Type Lined Cap (Smooth)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.