Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic F217 and PTFE Lined Caps

Catalog No. 02991112
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
38-430
38-439
70-400
Product Type:
Lined Cap (Ribbed)
Lined Cap (Smooth)
No. per Case:
144
576
840
2160
2800
3600
4400
6300
Packaging:
Bagged
Bulk
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type No. per Case Packaging
02-991-112 28-400 Lined Cap (Ribbed) 2800 Bulk
02-991-131 20-400 Lined Cap (Ribbed) 6300 Bulk
02-991-108 24-400 Lined Cap (Ribbed) 3600 Bulk
02-991-192 22-400 Lined Cap (Ribbed) 4400 Bulk
02-991-057 38-400 Lined Cap (Ribbed) 2160 Bulk
02-991-058 38-430 Lined Cap (Ribbed) 840 Bulk
02-991-020 38-439 Lined Cap (Ribbed) 840 Bulk
02-993-394 28-400 Lined Cap (Ribbed) 576 Bagged
02-993-373 24-400 Lined Cap (Ribbed) 576 Bagged
02-993-401 33-400 Lined Cap (Ribbed) 576 Bagged
02-993-432 70-400 Lined Cap (Smooth) 144 Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 02-991-112 Supplier Qorpak™ Supplier No. CAP00191
Add to Cart
Add to Cart
Request a Quote

Ideal for use with a wide range of chemicals including analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals and diagnostic reagents. Qorpak™ Black Phenolic F217 and PTFE Lined Caps offer good chemical and temperature resistance.

  • Adherence consistency to close dimensional tolerances
  • Resist from chemical attacks at room temperature
  • Provide resilient tight seal with low moisture transmission rate
  • Available in ribbed or smooth style
  • Available in bagged or bulk packaging

Specifications

Color Black
Material Phenolic Resin
No. per Case 2800
Packaging Bulk
Sterility Non-sterile
Liner Material F217, PTFE
Neck Finish 28-400
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.