Learn More
Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps
Description
- Not recommended for acids and alkalis
- Available as ribbed and smooth style
- Thread finish for narrow bottles is 425 GPI and for wider 400
- Available as bulk/bagged packaging
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 6500/Cs. |
| Packaging | Bulk |
| Liner Material | Pulp/Tin Foil |
| Neck Finish | 20-400 |
| Product Type | Lined Cap (Smooth) |
| For Use With (Equipment) | Wide mouth bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.