Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps

Catalog No. 02993327
Change view
Click to view available options
Neck Finish:
20-400
33-400
38-400
43-400
48-400
89-400
No. per Case:
1248/Cs.
144/Cs.
1680/Cs.
576/Cs.
6500/Cs.
Packaging:
Bagged
Bulk
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-993-327 33-400 576/Cs. Bagged
02-993-353 48-400 576/Cs. Bagged
02-991-104 38-400 1680/Cs. Bulk
02-991-036 48-400 1248/Cs. Bulk
02-993-361 89-400 144/Cs. Bagged
02-993-354 38-400 576/Cs. Bagged
02-993-352 43-400 576/Cs. Bagged
02-991-259 20-400 6500/Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 02-993-327 Supplier Qorpak™ Supplier No. CAP00215

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For use with solids and powders. Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps offer strong resistance to chemicals and temperature.

  • Not recommended for acids and alkalis
  • Available as ribbed and smooth style
  • Thread finish for narrow bottles is 425 GPI and for wider 400
  • Available as bulk/bagged packaging

Specifications

Color Black
Material Phenolic Resin
No. per Case 576/Cs.
No. per Pack 12/Pk.
Packaging Bagged
Liner Material Pulp/Tin Foil
Neck Finish 33-400
Product Type Lined Cap (Smooth)
For Use With (Equipment) Wide mouth bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.