Learn More
Grainger Black Pipe Nipple Threaded 1x12 In
Supplier:  Grainger 14J813
Encompass_Preferred
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from seamless carbon steel pipe. It has National Pipe Thread ends that comply with ANSI standards. The nipple has a pressure rating of 850 psi at 70°F. It can be used in steam, air, gas, oil, and water applications. The nipple's working temperature range is -20° to 800°F. It measures 6 in. long and has an outside diameter of 0.840 in. The nipple weighs 0.22 lbs.
Catalog No. 50-275-2497
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.