Learn More
MilliporeSigma™ Supelco™ Black Plastic Caps
Catalog No. 111009118 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
11-100-9118 18-400
11-100-9695 13-425
11-100-9592 15-425
11-100-9704 20-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-9118 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1888321PAK
Add to Cart
Add to Cart
Used as a closure for containers used to contain substances in order to prevent contamination, designed to ensure the ultimate in product safety
  • Solid top
  • PTFE-faced rubber liner
  • Prevents contamination
  • Easy sterilization
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE-Faced Rubber
Color Black
Material Phenolic
No. per Pack 200/Pk.
Closure Size 18-400
Autoclavable Autoclavable
Product Type Solid Top Cap