Learn More
MilliporeSigma™ Supelco™ Black Plastic Caps with White Rubber Liner
Shop All MilliporeSigma Supelco ProductsDescription
- Solid top
- White rubber liner
- Prevents contamination
- Easy sterilization
Specifications
Specifications
| Color (Seal) | White |
| Material (Liner) | Rubber |
| Color | Black |
| Material | Phenolic |
| No. per Pack | 500/Pk. |
| Closure Size | 20-400 |
| Autoclavable | Autoclavable |
| Product Type | Solid Top Cap |