Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Polypropylene Unlined Caps

Catalog No. 02993120
Change view
Click to view available options
For Use With (Equipment):
Wide mouth bottle
Neck Finish:
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
63-400
70-400
83-400
89-400
No. per Case:
1300 Cs.
1700 Cs.
2000 Cs.
2200 Cs.
2900 Cs.
4000 Cs.
500 Cs.
5000 Cs.
580 Cs.
6500 Cs.
760 Cs.
8500 Cs.
900 Cs.
Packaging:
Bagged
Bulk
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case Packaging
02-993-120 Wide mouth bottle 22-400 500 Cs. Bagged
02-991-121 Wide mouth bottle 89-400 580 Cs. Bulk
02-991-437 Wide mouth bottle 33-400 4000 Cs. Bulk
02-991-438 Wide mouth bottle 38-400 2900 Cs. Bulk
02-991-049 Wide mouth bottle 53-400 1300 Cs. Bulk
02-991-436 Wide mouth bottle 28-400 5000 Cs. Bulk
02-991-435 Wide mouth bottle 24-400 6500 Cs. Bulk
02-991-141 Wide mouth bottle 48-400 1700 Cs. Bulk
02-991-267 Wide mouth bottle 43-400 2200 Cs. Bulk
02-991-268 Wide mouth bottle 45-400 2000 Cs. Bulk
02-991-344 Wide mouth bottle 70-400 760 Cs. Bulk
02-991-266 Wide mouth bottle 22-400 8500 Cs. Bulk
02-991-175 Wide mouth bottle 83-400 500 Cs. Bulk
02-992-671 Wide mouth bottle 63-400 900 Cs. Bulk
02-993-174 Wide mouth bottle 28-400 500 Cs. Bagged
02-993-229 Wide mouth bottle 43-400 500 Cs. Bagged
02-992-703 Wide mouth bottle 24-400 500 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02-993-120 Supplier Qorpak™ Supplier No. CAP00333

Ideally used for dry products. Qorpak™ Black Polypropylene Unlined Caps are autoclavable.

  • Offer good impact strength and pliability
  • Available in ribbed style
  • Available as bulk/bagged packaging

Specifications

Color Black
Material Polypropylene
No. per Case 500 Cs.
No. per Pack 100/Pk.
Packaging Bagged
Sterility Non-sterile
Neck Finish 22-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Closure Type Ribbed
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.