Learn More
Labconco™ Bladder Tank Connect Kit
SDP
Catalog No. 16985639 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
16-985-639 Plastic
1 options
Catalog No. 16-985-639 Supplier Labconco™ Supplier No. 9201200
Add to Cart
Add to Cart
WaterPro Bladder Tanks offer additional storage of reverse osmosis water from an RO system.
  • Pressurized water is distributed to a polishing system, or glassware washer for multiple pure water rinses.
  • This kit connects multiple bladder tanks ensuring enough water for your process.
Specifications
Material Plastic
Product Type Water Connection