Learn More
R&D Systems™ Bladder Tissue Slides (Transitional Cell Carcinoma)- Paraffin
Shop All Novus Biologicals Products

Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Bladder Tissue Slides (Transitional Cell Carcinoma) |
| Regulatory Status | RUO |
| Species | Human |
| Tissue Condition | Transitional Cell Carcinoma |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.