Learn More
Thermco Products Inc BLANKET WARMER THERMOMETER EA
Supplier: Thermco Products Inc ACCG220F
Thermco's BI-Metal Dial Thermometers feature quality construction and design with easy to read black scale on a white background. The accuracy of these thermometers is +/- 1% over the entire scale. All stainless steel case, stem, and bezel for superior shock protection, even in extreme conditions. The lens is made from unbreakable polycarbonate. An external nut provides for easy recalibration. Immersion is approximately 2" (50mm). Supplied with a convenient plastic stem case which has a pocket clip. This case can also be used as a handle for the thermometer. 1 3/4in. dia x 8in. Stem, Lexan Lens, 0/220F x 2.0F
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.