Learn More
Invivogen Blasticidin (Solution), 100 mg (10 x 1 ml)
Supplier: Invivogen ANTBL1
Purity: ≥ 95% (HPLC), Product concentration: 10 mg/ml, Blasticidin is an efficient selection antibiotic that acts on both eukaryotic and prokaryotic cells. Blasticidin is a peptidyl nucleoside antibiotic isolated from Streptomyces griseochromogenes that inhibits protein synthesis by interfering with the peptide-bond formation in the ribosomal machinery.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.