Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blasticidin S Deaminase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143657 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143657 Supplier Invitrogen™ Supplier No. PA5143657
  • $631.50
    $537.65 / Each of 1

    Save $93.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The antibody detects a 13 kDa* protein corresponding to the molecular mass of Bsd on SDS-PAGE immunoblots of 293 cells transfected with a lentiviral vector that expresses the Bsd gene product, but is not detected in mock transfected cells.

Retroviral vectors are commonly used for gene expression in vitro and in vivo. The structure of these vectors usually consists of one gene of interest and a selectable marker gene. Fast selection without damaging cells is a critical step for significant gene expression. Blasticidin S deaminase (Bsd) isolated from Bacillus cereus has a neutralizing action on the highly toxic antibiotic blasticidin. A variety of commercially available retroviral vectors, such lentiviral expression systems, utilize Bsd expression for selection using blasticidin. This selectable marker allows selection of stable mammalian cell lines in as little as one week.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blasticidin S Deaminase
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene BAD
Gene Accession No. Q61337
Gene Alias blasticidin; BSD
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of Blasticidin S Deaminase* from Aspergillus Terreus.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12015
Target Species Fungi
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.