Learn More
Enzo Life Sciences Blasticidin S. hydrochloride (100mg). CAS: 3513-03-9
SDP
Supplier:  Enzo Life Sciences ALX380089M100
Encompass_Preferred
Antibiotic. Inhibitor of DNA and protein synthesis. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water; insoluble in organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-8950
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.