Learn More
Market Lab Bleach Rite Disinfecting Spray, 16oz
Supplier:  Market Lab 3004
Encompass_Preferred
Easy-to-Carry 16-Ounce Spray Bottle Makes Disinfecting Less of a Chore. Formulated at 0.525%, the 1:10 dilution recommended by CLSI, OSHA, and CDC. Registered with EPA. 18-month shelf life from date of manufacture (expiration date printed on each bottle). Pre-mixed and ready to use. Safe to use on plastic, ceramics, and glass.
Catalog No. NC9683648
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.