Learn More
STEMCELL Technologies (-)-Blebbistatin, Size: 25 mg
SDP
Supplier:  STEMCELL Technologies 72404
Encompass
(-)-Blebbistatin is a selective cell-permeable inhibitor of non-muscle myosin II ATPases (Kovács et al.; Straight et al.).  It rapidly and reversibly inhibits Mg-ATPase activity and in vitro motility of non-muscle myosin IIA and IIB for several species (IC₅₀ = 0.5-5.0 μM), while poorly inhibiting smooth muscle myosin (IC₅₀ = 80 μM) (Limouze et al.).MAINTENANCE AND SELF-RENEWAL· Increases human pluripotent stem cell (hPSC) survival and cloning efficiency after dissociation to single cells, downstream of ROCK inhibition (Chen et al.; Ohgushi et al.; Walker et al.; Xu et al.). · Enables hPSC to be cultured on microcarriers without surface coating (Chen et al.)· Inhibits differentiation of human mesenchymal stem cells (McBeath et al.; Engler et al.).
Catalog No. 50-197-5918
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.