Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bleomycin Sulfate, Anticancer and Antibiotic Agent, >98% Purity, Solid, 50 mg

Catalog No. 502583304
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3304 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3304 Supplier Abcam Supplier No. AB14297750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Radiomimetic anticancer and antibiotic agent that induces DNA damage and oxidative stress.

  • Molecular Weight: 1512.6 Da
  • Purity: >98%
  • Physical Form: Solid
  • Description: Radiomimetic anticancer and antibiotic agent that induces oxidative stress and DNA damage via single strand breaks, double strand breaks, and apurinic/apyrimidinic modifications. Shows anticancer effects in vivo.
  • Storage: Store at +4°C.
  • Citation Count: 9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule DNA
Shipping Condition Ambient
Buffer 98% Bleomycin sulfate
Alternate Part Number Blenoxane
Content And Storage Store at 4°C.
Form Solid
Formulation 98% Bleomycin sulfate
Quantity 50 mg
For Use With (Application) Anticancer research, antibiotic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.