Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bleomycin Sulfate, MP Biomedicals™

Catalog No. MP219030610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219030610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219030610 Supplier MP Biomedicals, Inc Supplier No. 0219030610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bleomycin Sulfate from Streptomyces verticillus, White powder

  • It is an atineoplastic antibiotic.
  • A mixture of cytotonic glycopeptide antibiotics which reacts with DNA and causes strand scission; they have also been shown to have a type of oxygen transferase activity. Bleomycin A2 is the main component of the bleomycin employed clinically.
  • Bleomycins differ from one another in the terminal amine and show varying biological activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-93-4
Molecular Formula C55H85N17O25S4
Molecular Weight (g/mol) 1512.62
MDL Number MFCD00070310
InChI Key WUIABRMSWOKTOF-OCBSMOPSSA-N
Synonym Blenoxane, Blexane, Bleo
PubChem CID 122172957

Specifications

Color White
Quantity 10 mg
Solubility Information Very soluble in water, methanol; slightly soluble in ethanol. Practically insoluble in acetone, ethyl acetate, butyl acetate, ether.
Grade >99.0%
Physical Form Powder
RTECSNumber EC5991990
storageNote1 Store at 2-8°C,Store Desiccated & protect from light
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.