Learn More
Selleck Chemical LLC Bleomycin Sulfate 10mg 9041-93-4 NSC125066
SDP
Supplier:  Selleck Chemical LLC S121410MG
Encompass_Preferred
Bleomycin Sulfate (NSC125066) is a glycopeptide antibiotic and an anticancer agent for squamous cell carcinomas (SCC) with IC50 of 4 nM in UT-SCC-19A cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-673-3
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.