Learn More
Invitrogen™ BLNK Recombinant Rabbit Monoclonal Antibody (JE63-50)

Description
BLNK Recombinant Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | BLNK |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE63-50 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BLNK |
| Gene Accession No. | Q4KM52, Q8WV28, Q9QUN3 |
| Gene Alias | AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.