Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Block Heater

Catalog No. 111006933 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Voltage:
110 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
11-100-6933 220 V
11-100-8015 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-6933 Supplier MilliporeSigma™ Supelco™ Supplier No. 33318U
Add to Cart
Add to Cart

Heating Blocks Sold Separately

  • Multi-purpose unit ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
  • Accepts separate, interchangeable modular heating blocks which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
  • Two Temperature Adjustments: Ambient to 100°C (Low Range) or 75° to 150°C (High Range)
  • Blocks include a thermometer well for measuring block temperature
  • Economical, versatile and compact
  • Heating Blocks sold separately
TRUSTED_SUSTAINABILITY

Specifications

Description Block Not Included
Plug Type EU
Temperature Range (Metric) Ambient to 150°C
For Use With (Application) Incubation/Activation of Cultures, Enzyme Reactions, Immunoassays, Melting/Boiling Points, and More
Heat Up Time 45 minutes (to 100°C)
Includes Thermometer Well
Temperature Stability ±0.1°C (at 37°C)
Temperature Uniformity ±0.1°C (at 37°C)
Dimensions (L x W x H) 12.4 x 8 x 3.5 in. (31.5 x 20.3 x 8.9 cm)
Shipping Weight (English) 5.8 lb.
Shipping Weight (Metric) 2.6 kg
Voltage 220 V
Wattage 110 watts
Amperage 0.5 amps
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.