Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate

Catalog No. R02050
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
85mm biplate
Media Type:
TSA-5% Sheep Blood/MacConkey
Manufacturing Origin:
United States
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R02050 10/Pk. 85mm biplate TSA-5% Sheep Blood/MacConkey United States
R02049 100/Pk. 85mm biplate TSA-5% Sheep Blood/MacConkey United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R02050 Supplier Thermo Scientific™ Supplier No. R02050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Grow fastidious organisms, detect and enumerate coliforms, and isolate and detect Salmonella and Shigella spp. on a single plate.

Isolate and cultivate fastidious microorganisms with clearly visible hemolytic reactions (staphylococci and streptococci) and detect and enumerate coliform organisms such as E. coli, while at the same time isolating and detecting Salmonella and Shigella spp., using Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media. One half of the plate contains TSA with 5% sheep blood to promote growth of fastidious organisms with clearly visible hemolysis forms for staphylococci and streptococci. MacConkey Agar on the other half of the plate is a selective medium for differentiating between coliforms and lactose and non-lactose fermenters.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Opaque red/ clear pink
Content And Storage 2°C to 8°C
Format 85mm biplate
Manufacturing Origin United States
Media Type TSA-5% Sheep Blood/MacConkey
Product Type Agar Plate
Fill Volume 10 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.